Learn More
VACUUBRAND™ Stainless Steel Vacuum Tubing With Small Flanges KF
Description
- Manufactured from stainless steel
- Flange and tube TIG welded
- Available in a range of sizes
Specifications
Specifications
| Material | Stainless Steel |
| Diameter (Metric) Outer | 25 mm |
| Packaging Quantity | 1 Each |
| Tubing Size | 100mm |
| Includes | Small flanges |
| Product Type | Vacuum Tubing |