Learn More
Erlab Stainless Steel Worksurface Captair 633 Smart
Product Code. 15613784 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Captair 633 Ductless Filtering Fume Hood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15613784 Captair 633 Ductless Filtering Fume Hood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15613784 Supplier Erlab Supplier No. I0165050001
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Worksurface for your ductless fume hoods.
  • Worksurface for your ductless fume hood Captair 633 Smart
  • Material: Stainless steel
  • Equipment: Spill rentention tray
Specifications
For Use With (Equipment) Captair 633 Ductless Filtering Fume Hood
Product Type Work Surface