Learn More
Rogo Sampaic™ Stand
Product Code. 11702674
Change view
Click to view available options
Size:
180 mm
120 x 210 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11702674 120 x 210 mm Each
11722674 180 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11702674 Supplier Rogo Sampaic™ Supplier No. FFB515
Specifications
Product Type Stand
Length (Metric) 210 mm
Size 120 x 210 mm
Dimensions (L x W) 210 x 120 mm
Height (English) 3 ft.
No. per Pack 1 Ea.
Width (Metric) 120 mm