Learn More
Waring™ Standard Blender
Product Code. 11942919
Change view
Click to view available options
1 Speed
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11942919 1 Speed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 11942919 Supplier Waring™ Supplier No. 800ES
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Specifications
Amperage 1.8 A
Speed 22,000 rpm
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz, 1.8A
Capacity (Metric) 1 mL
Packaging Quantity 1 Ea.