Learn More
Bochem™ Standard Retort Clamp
Bochem™ Standard Retort Clamp with a round, cork lined jaw.
Brand: Bochem™ 5451
Additional Details : Poids : 0.50000kg
Description
- DIN 12894
- Cork lining
Specifications
Stainless Steel (18/10) | |
180 mm | |
Round |
12 mm | |
120 mm |