Learn More
Invitrogen™ Staphylococcus aureus Monoclonal Antibody (Staph11-248.2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46115
Description
MA1-10709 detects Staphylococcus aureus from Staph. Aureus and Staph. epidermidis samples. MA1-10709 has been successfully used in ELISA applications. The MA1-10709 immunogen is UV-inactivated Staphylococcus aureus cells.
Staphylococcus aureus is a type of bacteria that commonly lives on the skin and in the nasal passages of healthy individuals. However, it can also cause a range of infections, from minor skin infections to more serious conditions such as pneumonia, sepsis, and endocarditis.
Specifications
| Staphylococcus aureus | |
| Monoclonal | |
| 2.2 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgM |
| ELISA | |
| Staph11-248.2 | |
| Unconjugated | |
| MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant | |
| UV-inactivated Staphylococcus aureus cells. | |
| 1 mg | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.