missing translation for 'onlineSavingsMsg'
Learn More

Starch 5 g/L in Water Indicator Solution For Iodometry, Chem Lab

Product Code. 11903353
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11903353 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11903353 Supplier Chem-Lab Supplier No. CL06.2601.0250
  • ONLINE EXCLUSIVE PRICE
    34.00€
    17.20€ / 250mL
    Save 16.80€
    49% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 9005-84-9
Boiling Point 100°C
Melting Point 0°C
Concentration 5 g/L
Density 1 g/mL
Packaging Polyethylene
Quantity 250 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.