missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15689-1-AP-150UL
This item is not returnable.
View return policy
Description
Although the function of this gene is not known, its existence is supported by mRNA and EST data. The predicted gene product contains a region similar to the STAR-related lipid transfer (START) domain, which is often present in proteins involved in the cell signaling mediated by lipid binding. Alternatively spliced transcript variants have been described, although some transcripts occur only in cancer cell lines.Specifications
| STARD7 | |
| Polyclonal | |
| Unconjugated | |
| STARD7 | |
| GTT1, STARD7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296128, 56910, 99138 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R1R3, Q9NQZ5 | |
| STARD7 | |
| STARD7 Fusion Protein Ag8224 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction