missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Antibody (CL0492), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52936-25ul
This item is not returnable.
View return policy
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
| STAT3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| CL0492 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GVTFTWVEKDISGKTQIQSVEPYTKQQLNNMSFAEIIMGYKIMDATNILVSPLVYLYPDIPKEEAFGKYCRPESQEHPEADPGSAAPYLKTKFICVTPTTCSNTIDLPMSPRTLDSLMQFGNNGEGAEPSAGGQFESLTFDMELTSECA | |
| 25 μL | |
| Cellular Markers, Myeloid derived Suppressor Cell, Signal Transduction, Transcription Factors and Regulators | |
| 6774 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction