missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STAT5 beta Recombinant Rabbit Monoclonal Antibody (24GB2110)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00€ - 564.00€

Specifications

Antigen STAT5 beta
Clone 24GB2110
Concentration 0.53 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276125
GREENER_CHOICE
View Documents
Invitrogen™
MA557740
100 μL
564.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275140
View Documents
Invitrogen™
MA557741
20 μL
198.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

STAT5 beta Recombinant Monoclonal Antibody for Western Blot, Flow

The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.
Specifications

Specifications

STAT5 beta
0.53 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P42232, P51692, P52632
20851, 25126, 6777
A synthesized peptide derived from human STAT5B (aa 500-787).
Antibody
24GB2110
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
Stat5b
LOC101122961; mammary gland factor STAT5B; Signal transducer and activator of transcription 5a; signal transducer and activator of transcription 5B; STA5B; STAT; STAT5; STAT5B; transcription factor STAT5B
Stat5b
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.