missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT5B Mouse anti-Human, Clone: 1D9B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66427-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the STAT family of transcription factors. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein mediates the signal transduction triggered by various cell ligands, such as IL2, IL4, CSF1, and different growth hormones. It has been shown to be involved in diverse biological processes, such as TCR signaling, apoptosis, adult mammary gland development, and sexual dimorphism of liver gene expression. This gene was found to fuse to retinoic acid receptor-alpha (RARA) gene in a small subset of acute promyelocytic leukemias (APLL). The dysregulation of the signaling pathways mediated by this protein may be the cause of the APLL.Specifications
| STAT5B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51692 | |
| STAT5B | |
| STAT5B Fusion Protein Ag2709 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1D9B11 | |
| Unconjugated | |
| STAT5B | |
| STAT5, STAT5B | |
| Mouse | |
| Protein A | |
| RUO | |
| 6777 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction