missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT6 Mouse anti-Human, Mouse, Rat, Clone: 2E5B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66717-1-IG-20UL
This item is not returnable.
View return policy
Description
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.Specifications
| STAT6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P42226, P52633 | |
| Stat6 | |
| STAT6 Fusion Protein Ag0410 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 2E5B8 | |
| Unconjugated | |
| STAT6 | |
| D12S1644, IL 4 STAT, STAT6, STAT6B, STAT6C | |
| Mouse | |
| Protein A | |
| RUO | |
| 20852, 362896, 6778 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction