missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stathmin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11157-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the stathmin family of genes. It encodes a ubiquitous cytosolic phosphoprotein proposed to function as an intracellular relay integrating regulatory signals of the cellular environment. The encoded protein is involved in the regulation of the microtubule filament system by destabilizing microtubules. It prevents assembly and promotes disassembly of microtubules. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| Stathmin 1 | |
| Polyclonal | |
| Unconjugated | |
| STMN1 | |
| C1orf215, Lag, LAP18, Metablastin, Oncoprotein 18, OP18, Phosphoprotein p19, PP17, PP19, PR22, Prosolin, Protein Pr22, SMN, Stathmin, stathmin 1/oncoprotein 18, Stathmin1, STMN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16765, 29332, 3925 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P13668, P16949, P54227 | |
| Stmn1 | |
| Stathmin 1 Fusion Protein Ag1633 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction