Learn More
Mecmesin™ Static Torque Screwdriver
Product Code. 11957999
Encompass
Change view
Click to view available options
Capacity (Metric):
1.5 Nm
6 Nm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11957999 6 Nm Each
11937999 1.5 Nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11957999 Supplier Mecmesin™ Supplier No. 871005
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Suitable for the measurement of very fine static torque.
  • A range of fine-precision, low-capacity torque screwdrivers for handheld applications.
  • Equipped with 1/4 in. hex drive (or 3mm bore) for the fitting of adaptors.
  • Accuracy ±0.5% of the full scale
Tekniske data
Capacity (Metric) 6 Nm
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.