missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Mesoderm Induction Medium

Product Code. 17128261
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17128261 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17128261 Supplier STEMCELL Technologies Supplier No. 05221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete medium for the induction of early mesodermal differentiation

  • STEMdiff™ Mesoderm Induction Medium (MIM) is a defined, xeno-free medium for generation of early mesoderm cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells
  • Protocols for mesodermal differentiation can be difficult and inconsistent, therefore, use the short and simple STEMdiff™ MIM monolayer protocol to differentiate your human pluripotent stem cells (hPSCs)
  • STEMdiff™ MIM is a complete medium that produces a cell population enriched for early mesoderm, as indicated by positive expression of Brachyury (T) and NCAM markers
  • As part of the hPSC workflow, STEMdiff™ MIM efficiently differentiates hPSCs cultured in TeSR™ media
  • When directed, early mesoderm cells produced using STEMdiff™ MIM can be further differentiated to specialized cell types, such as osteoblasts, chondrocytes, adipocytes or endothelial cells
  • For more information, see the data below

Specifications

Cell Type Mesoderm, PSC-Derived, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.