Learn More
STEMCELL Technologies STEMdiff™ Neural Induction Medium
Product Code. 17148301
Encompass
Change view
Click to view available options
Quantity:
250 mL
2 x 250 mL
Unit Size:
250mL
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17148301 250 mL 250mL
17158301 2 x 250 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17148301 Supplier STEMCELL Technologies Supplier No. 05835
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Defined, serum-free medium for neural induction of human ES and iPS cells
  • STEMdiff™ Neural Induction Medium is a defined, serum-free medium for the neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • This medium enables highly efficient generation of neural progenitor cell using either embryoid body- or monolayer culture-based protocols
Specifications
Cell Type Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 250 mL