Learn More
STEMCELL Technologies STEMdiff™ Neural Progenitor Freezing Medium
Product Code. 15385811
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15385811 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15385811 Supplier STEMCELL Technologies Supplier No. 05838
For cryopreservation of neural progenitor cells generated using STEMdiff™ Neural Induction Medium
  • STEMdiff™ Neural Progenitor Freezing Medium is a serum-free medium for cryopreservation of neural progenitor cells (NPCs) derived from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
  • This freezing medium is optimized for the cryopreservation of NPCs generated using STEMdiff™ Neural Induction Medium (Catalog #05835) and cultured in STEMdiff™ Neural Progenitor Medium (Catalog #05833)
  • NPCs can be frozen at any point post-neural induction, with reproducibly high recovery rates
  • Post-thaw, NPCs display healthy morphology, express NPC markers, and retain the potential to expand and differentiate into neurons
Specifications
Cell Type Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 100 mL