Learn More
Zeiss™ Stemi 305 Trinocular Stereo Microscope
Product Code. 17100686
Change view
Click to view available options
Includes:
Double Spot Illuminator K LED
Ring Illuminator K LED
Ring Illuminator VisiLED
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
17100686 Double Spot Illuminator K LED Each
17322830 Ring Illuminator K LED 1 set
17342830 Ring Illuminator VisiLED 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is under eco-contribution tax.
Product Code. 17100686 Supplier Zeiss™ Supplier No. 4919030006000
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Includes Double Spot Illuminator K LED
Design Type Trinocular