missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemRegenin 1 (Hydrochloride)

Product Code. p-7217572
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17119251 1 mg 1mg
17129251 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17119251 Supplier STEMCELL Technologies Supplier No. 72352
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aryl hydrocarbon receptor (AHR) antagonist

  • StemRegenin1 (SR1) is an antagonist of the aryl hydrocarbon receptor (AHR)
  • It promotes ex vivo expansion of CD34+ human hematopoietic stem cells (Boitano et al.), 2010, Csazar et al.), 2012)
  • SR1 has been shown to collaborate with UM729 in preventing differentiation of acute myeloid leukemia (AML) cells in culture (Pabst et al.), 2014)
  • SR1 also stimulates the proliferation and differentiation of CD34+ hematopoietic progenitor cells into dendritic cells (Thordardottir et al.), 2014)
  • This product is supplied as the hydrochloride salt of the molecule

Specifications

Synonym SR1 (Hydrochloride)
Quantity 1 mg
Molecular Formula C24H23N5OS · HCl
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.