missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemSpan™-ACF Without Phenol Red
Description
- StemSpan™-ACF Without Phenol Red is an animal component-free (ACF) medium that has been developed for the in vitro culture and expansion of human hematopoietic cells, when appropriate growth factors/cytokines and supplements (e.g. small molecules) are added
- This allows users the flexibility to prepare medium that meets their requirements
- StemSpan™-ACF Without Phenol Red contains only recombinant proteins and synthetic components, and does not contain serum or other human- or animal-derived components
- Using appropriate cytokines, StemSpan™-ACF Without Phenol Red may be used to expand CD34+ cells isolated from human cord blood, mobilized peripheral blood, or bone marrow samples, or to expand and differentiate lineage-committed progenitor cells to generate populations of myeloid or megakaryocyte progenitor cells
- StemSpan™-ACF Without Phenol Red is manufactured and tested following relevant cGMPs under a certified quality management system
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Differentiation, Expansion, Genome Editing |
| Quantity | 500 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?