missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemSpan™ Lymphoid Differentiation Coating Material (100X)

Product Code. 17159961
Encompass
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17159961 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17159961 Supplier STEMCELL Technologies Supplier No. 09925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Coating material for the differentiation of lymphoid cells

  • StemSpan™ NK Cell Generation Kit has been developed to differentiate CD34+ cells isolated from cord blood (CB) and bone marrow (BM) to natural killer (NK) cells, without use of stromal cells
  • This kit typically promotes the expansion of thousands of CD56+ NK cells in cultures initiated with CD34+ human CB cells
  • StemSpan™ Lymphoid Progenitor Expansion Supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion and differentiation of CD34+ cells isolated from human CB and BM samples to lymphoid progenitor cells when used in combination with StemSpan™ SFEM II medium, and on plates coated with StemSpan™ Lymphoid Differentiation Coating Material (100X)
  • Subsequently, StemSpan™ NK Cell Differentiation Supplement (100X) enables differentiation of lymphoid progenitor cells to NK cells

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, NK Cells
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 0.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.