missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sterigmatocystin Monoclonal Antibody (4D3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549620
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Sterigmatocystin is a toxic secondary metabolite produced by certain species of fungi, including Aspergillus versicolor and Aspergillus nidulans. It is closely related to aflatoxin, another fungal toxin, and has been shown to have carcinogenic and mutagenic properties. Sterigmatocystin is commonly found in contaminated grains and nuts, and can also be present in feed for livestock.
Specifications
| Sterigmatocystin | |
| Monoclonal | |
| 7.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 4D3 | |
| Unconjugated | |
| Sterigmatocystine | |
| Conjugate of STG hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction