Learn More
Kartell™ Sterile Graduated Conical Centrifuge Tube
Product Code. 16677363
Change view
Click to view available options
Capacity (Metric):
15 mL
50 mL
Graduations:
0.5 mL
2.5 mL
Max. RCF:
6200 RCF
9400 RCF
Packaging Type:
Bulk Pack
Single Pack
Tray Pack
Unit Size:
Pack of 1000
Pack of 500
Pack of 750
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Max. RCF Packaging Type unitSize
16677363 15 mL 0.5 mL 6200 RCF Tray Pack Pack of 750
16657363 15 mL 0.5 mL 6200 RCF Bulk Pack Pack of 1000
16667363 15 mL 0.5 mL 6200 RCF Single Pack Pack of 750
12553347 50 mL 2.5 mL 9400 RCF Tray Pack Pack of 500
12533347 50 mL 2.5 mL 9400 RCF Bulk Pack Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16677363 Supplier Kartell™ Supplier No. 8401000
Only null left
Add to basket
Add to basket
Increase sample recovery with a conical shaped tube that features a ribbed, easy to use cap. Kartell™ Sterile Graduated Conical Centrifuge Tube is made of high transparency polypropylene and has a large white marking area to increase traceability.
  • Withstands temperature extremes of -80°C to +121°C
  • Includes blue thread-design plug-seal cap with ribbed edge for safe and easy opening and closing
  • Conical bottom for total sample recovery
  • Easy-to-read graduations are pre-printed
  • Designed for sample collection or transportation
  • Apyrogenic and endotoxines free
  • Tubes are irradiated by ionizing beta radiation
Specifications
Closure Color Blue
Diameter (Metric) 17 mm
Max. RCF 6200 RCF
Autoclavable Autoclavable
Certifications/Compliance CE/IVD
Closure Type Plug Seal
Color Crystal Clear
Length (Metric) 121 mm
Material Polypropylene
Graduated Yes
Includes White marking area
Packaging Type Tray Pack
Sterility Sterile
Capacity (Metric) 15 mL
Graduations 0.5 mL
Shape Conical
Show More Show Less
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.