Description
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
- BSE/TSE free
- Full batch traceability
- Suitable for use in ATEX areas
- Available pre-sterilized (SAL 10-6)
Specifications
Specifications
| Material | HDPE |
| Sterility | Sterile |
| Length (Metric) | 400 mm |
| Width (Metric) | 25 mm |