missing translation for 'onlineSavingsMsg'
Learn More

STF 083010, TRC

Product Code. 30423889
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423889 10 mg 10mg
30410073 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30423889 Supplier Toronto Research Chemicals Supplier No. TRCS68674010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307543-71-1
Molecular Formula C15H11NO3S2
Molecular Weight (g/mol) 317.38
Synonym STF 083010, N-[(2-Hydroxy-1-naphthalenyl)methylene]-2-thiophenesulfonamide
IUPAC Name (NE)-N-[(2-hydroxynaphthalen-1-yl)methylidene]thiophene-2-sulfonamide
SMILES Oc1ccc2ccccc2c1\C=N\S(=O)(=O)c3cccs3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H11NO3S2/c17-14-8-7-11-4-1-2-5-12(11)13(14)10-16-21(18,19)15-6-3-9-20-15/h1-10,17H/b16-10+
Formula Weight 317.018
Chemical Name or Material N-[(2-Hydroxy-1-naphthalenyl)methylene]-2-thiophenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.