missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STIL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565446
This item is not returnable.
View return policy
Description
Immunogen sequence: RLLEAQSLMPC SPKTTAVEDT VQAGRQMELV SVEAQSSPGL HMRKGVSIAV STGASLFWNA AGEDQEPDSQ MKQDDTKI Highest antigen sequence identity to the following orthologs - mouse 85%, rat 81%.
Plays an essential role in early embryonic development.
Specifications
| STIL | |
| Polyclonal | |
| Unconjugated | |
| STIL | |
| cassiopeia; chunp6907; csp; MCPH7; nbb; scl/tal1 interrupting locus; SCL-interrupting locus protein; SCL-interrupting locus protein homolog; SIL; sill; Stil; STIL centriolar assembly protein; stil protein; TAL1 (SCL) interrupting locus like; Tal1 interrupting locus; TAL1/SCL interrupting locus-like protein; TAL-1-interrupting locus protein; zgc:110502 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6491 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15468 | |
| STIL | |
| Recombinant Human STIL. Recombinant protein control fragment (Product #RP-104823). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction