Learn More
OHAUS™ Stirrer Shaft
Product Code. 17094227 Shop All Ohaus Products
Change view
Click to view available options
Shape:
Anchor Blade
Fixed Blade
Floating Blade
Folding Blade
Paddle 6 Holdes
Propeller
Turbine
Turbo Propeller
Unit Size:
1 unit
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Shape Unit Size
17094227 Turbine 1 unit
17034227 Anchor Blade 1 unit
17044227 Fixed Blade 1 unit
17054227 Floating Blade 1 unit
17064227 Folding Blade 1 unit
17074227 Paddle 6 Holdes 1 unit
17084227 Propeller 1 unit
17004237 Turbo Propeller 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17094227 Supplier OHAUS™ Supplier No. 30586781
In Stock
Add to basket
Add to basket
Stirrer Shaft for use with overhead stirrers.
  • Overhead Stirrer Accessory
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 6.000 cm
Shape Turbine
Width (Metric) 10.000 cm
Weight (Metric) 200.000 g
Dimensions (L x W x H) 53.000 x 10.000 x 6.000 cm