Learn More
STK4/MST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22245-1-AP-20UL
 View more versions of this product
Product Code. 16871785
  • 145.00€ / 20µL
Description
Stress-activated, pro-apoptotic kinase which, following caspase-cleavage, enters the nucleus and induces chromatin condensation followed by internucleosomal DNA fragmentation. Key component of the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. STK3/MST2 and STK4/MST1 are required to repress proliferation of mature hepatocytes, to prevent activation of facultative adult liver stem cells (oval cells), and to inhibit tumor formation.
Specifications
Specifications
STK4/MST1
Polyclonal
Unconjugated
STK4
KRS2, MST 1, MST1, serine/threonine kinase 4, STE20 like kinase MST1, STK4, YSK3
Rabbit
Antigen Affinity Chromatography
RUO
311622, 58231, 6789
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.56 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13043, Q9JI11
Stk4
STK4/MST1 Fusion Protein Ag17738
20 μL
Primary
Human, Mouse, Rat