missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRADB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10688-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that belongs to the serine/threonine protein kinase STE20 subfamily. One of the active site residues in the protein kinase domain of this protein is altered, and it is thus a pseudokinase. This protein is a component of a complex involved in the activation of serine/threonine kinase 11, a master kinase that regulates cell polarity and energy-generating metabolism. This complex regulates the relocation of this kinase from the nucleus to the cytoplasm, and it is essential for G1 cell cycle arrest mediated by this kinase. The protein encoded by this gene can also interact with the X chromosome-linked inhibitor of apoptosis protein, and this interaction enhances the anti-apoptotic activity of this protein via the JNK1 signal transduction pathway. Two pseudogenes, located on chromosomes 1 and 7, have been found for this gene.Specifications
| STRADB | |
| Polyclonal | |
| Unconjugated | |
| STRADB | |
| ALS2CR2, CALS 21, ILP interacting protein, ILPIP, ILPIPA, PAPK, PRO1038, Pseudokinase ALS2CR2, STRAD beta, STRADB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 227154, 501146, 55437 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K4T3, Q9C0K7 | |
| STRADB | |
| STRADB Fusion Protein Ag1040 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction