Description
- Range of different convenient lengths
Specifications
Specifications
| Product Type | Tweezers |
| Length (English) | 15.7 in. |
| Length (Metric) | 400 mm |
| Material | Stainless Steel (316L) |
| Quantity | 1 Tweezer/Cs. |
| Sterility | Non-sterile |
| Style | Standard |
| Tip Style | Straight |