Learn More
IBA Lifesciences Strep-Tactin™XT 4Flow™ High Capacity Column
Product Code. 16614734 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
5 x 0.2 mL
Unit Size:
1 set
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16614734 1 mL 1 set
16604734 5 x 0.2 mL 1 set
16624734 5 mL 1 set
16634734 10 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16614734 Supplier IBA Lifesciences Supplier No. 25032001
  • ONLINE EXCLUSIVE PRICE
    148.00€
    122.00€ / 1 sets
    Save 26.00€
    17% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT 4Flow™ high capacity columns contain a 4% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
  • Due to the broad pH compatibility, excellent pressure stability, outstanding target purity (> 95%), and highest binding affinity, the resin is recommended for all protein classes, especially for low abundant or challenging proteins.
  • The low concentrated agarose enables the simple attachment of larger proteins to the stationary phase, making it particularly suitable for purification of high-molecular-weight targets.
  • In addition, Strep-Tactin™XT 4Flow™ high capacity allows the application of denaturing agents such as urea.
  • In comparison to Strep-Tactin™XT 4Flow™, Strep-Tactin™XT 4Flow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 2-fold higher yield of purified protein.
  • To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Matrix 4% agarose, crosslinked
Particle Size 50 to 150 μm
Quantity 1 mL
Target Twin-Strep-Tag™ and Strep-tag™II
Format Pre-packed Gravity Flow Column