missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Streptococcus agalactiae Monoclonal Antibody (Strep12-504.5)
GREENER_CHOICE

Product Code. 11543463
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11543463 500 μg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11543463 Supplier Invitrogen™ Supplier No. MA110874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10874 detects Streptococcus agalactiae from Strep agalactiae samples. MA1-10874 has been successfully used in ELISA applications. The MA1-10874 immunogen is UV-inactivated Strep agalactiae cells isolated from mastitic milk.

Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus agalactiae
Applications ELISA
Classification Monoclonal
Clone Strep12-504.5
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias s agalactiae; S. agalactiae
Host Species Mouse
Immunogen UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.