missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Striated Muscle Myosin Heavy Chain Antibody (83B6), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50254APC
This item is not returnable.
View return policy
Description
Striated Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Striated Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Striated Muscle Myosin Heavy Chain | |
| Monoclonal | |
| APC | |
| PBS | |
| Purified human skeletal muscle myosin | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 83B6 | |
| Western Blot, Immunohistochemistry | |
| Mouse | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Chicken, Rabbit | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction