Learn More
Cole-Parmer™ Stuart™ Flat Lid
Product Code. 12737969
Change view
Click to view available options
For Use With (Equipment):
12L Bath
18 and 26L Bath
48L Bath
8L Bath
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12737969 48L Bath Each
12717969 12L Bath Each
12727969 18 and 26L Bath Each
12707969 8L Bath Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12737969 Supplier Cole-Parmer™ Supplier No. 1212294
  • ONLINE EXCLUSIVE PRICE
    260.00€
    222.91€ / Pack of 1
    Save 37.09€
    14% Off
Estimated Shipment: 09-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enhance the performance of your Techne baths and circulators
  • Lids, trays, temperature probes, and other accessories for your bath.
  • Manufactured from stainless steel and available to fit 8 L baths to help prevent evaporation losses.
Specifications
For Use With (Equipment) 48L Bath