missing translation for 'onlineSavingsMsg'
Learn More

Styrene (stabilized with 4-tert-Butylcatechol), TRC

Product Code. 30138583
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138583 25 g 25g
30137322 5 g 5g
30040834 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138583 Supplier Toronto Research Chemicals Supplier No. TRCS68779025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8 H8
Molecular Weight (g/mol) 104.15
Synonym Styrene, Benzene, ethenyl-, Styrene (8CI), Ethenylbenzene, BYX 6, Cinnamene, EM 50, Maomin SM, NSC 62785, Phenethylene, Phenylethene, Phenylethylene, S 0095, STC, Styrol, Styrole, Styrolene, Styrolution PS 124N, Styropol SO, TTB 7302, Vinylbenzene, Vinylbenzol
IUPAC Name styrene
SMILES C=Cc1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2
Formula Weight 104.0626
Percent Purity >95%
Chemical Name or Material Styrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.