missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUB1 Antibody (04), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06541-100ul
This item is not returnable.
View return policy
Description
SUB1 Monoclonal antibody specifically detects SUB1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| SUB1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| 04 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:20-1:100, Immunoprecipitation 1-4 μg/mg of lysate | |
| activated RNA polymerase II transcription cofactor 4, activated RNA polymerase II transcriptional coactivator p15, MGC102747, p15, PC4p14P15, Positive cofactor 4, RPO2TC1, SUB1 homolog, SUB1 homolog (S. cerevisiae) | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SUB1 (Uniprot#: P53999; Met1-Leu127). | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 10923 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction