missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Succinimidyl acetate

Product Code. 16554284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16554284 100 g 100g
16534284 5 g 5g
16544284 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16554284 Supplier Thermo Scientific Chemicals Supplier No. 464751000
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 14464-29-0 | C6H7NO4 | 157.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14464-29-0
Molecular Formula C6H7NO4
Molecular Weight (g/mol) 157.13
InChI Key SIFCHNIAAPMMKG-UHFFFAOYSA-N
IUPAC Name 2,5-dioxopyrrolidin-1-yl acetate
SMILES CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 133.0°C to 134.0°C
Quantity 100 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in toluene
Formula Weight 157.13
Chemical Name or Material Succinimidyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.