missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Succinyl-HIST1H2AG (Lys95) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121472
This item is not returnable.
View return policy
Description
Antibody tested in Indirect ELISA.
Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
Specifications
| Succinyl-HIST1H2AG (Lys95) | |
| Polyclonal | |
| Unconjugated | |
| H2AC11 | |
| H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8329, 8969 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0C0S8 | |
| H2AC11, HIST1H2AG | |
| Synthesized peptide derived from Human Histone H2A type 1 protein (86-98aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction