Learn More
Succinyl sulfathiazole, 99.48%, MP Biomedicals™
Product Code. 11413351 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413351 100 g 100g
11423351 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11413351 Supplier MP Biomedicals Supplier No. 0210297680
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
Succinylsulfathiazole is used to block the synthesis of folic acid. It is used to study folate deficiency
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 116-43-8
Molecular Formula C13H13N3O5S2
Molecular Weight (g/mol) 355.383
MDL Number MFCD00022437
InChI Key SKVLYVHULOWXTD-UHFFFAOYSA-N
Synonym succinylsulfathiazole, cremosuxidine, sulfasuccidine, sulfasuccinil, sulfasuxidine, colistatin, sulfadigesin, sulfasuccithiazole, sulfasuccidin, sulfenterone
PubChem CID 5315
ChEBI CHEBI:9309
IUPAC Name 4-oxo-4-[4-(1,3-thiazol-2-ylsulfamoyl)anilino]butanoic acid
SMILES C1=CC(=CC=C1NC(=O)CCC(=O)O)S(=O)(=O)NC2=NC=CS2
Specifications
Color White
Quantity 100 g
Formula Weight 355.4
Physical Form Powder
Chemical Name or Material Succinyl sulfathiazole