missing translation for 'onlineSavingsMsg'
Learn More

Succinylsulfathiazole, TRC

Product Code. 30420491
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420491 10 g -
30417553 50 g 50g
30410275 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420491 Supplier Toronto Research Chemicals Supplier No. TRCS69217010G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-43-8
Molecular Formula C13 H13 N3 O5 S2
Molecular Weight (g/mol) 355.39
Synonym Butanoic acid, 4-oxo-4-[[4-[(2-thiazolylamino)sulfonyl]phenyl]amino]-, Succinanilic acid, 4'-(2-thiazolylsulfamoyl)- (7CI,8CI), 2-(N4-Succinylsulfanilamido)thiazole, 4'-(2-Thiazolylsulfamoyl)succinanilic acid, Colistatin, Cremosuxidine, Kaoxidin, Kaoxidine, NSC 14193, NSC 163939, Rolsul, Succinylsulfathiazole, Succinylsulphathiazole, Sulfadigesin, Sulfasuccidin, Sulfasuccidine, Sulfasuccinil, Sulfasuccithiazole, Sulfasuxidine, Sulfenterone, Thiacyl, p-2-Thiazolylsulfamylsuccinanilic acid
IUPAC Name 4-oxo-4-[4-(1,3-thiazol-2-ylsulfamoyl)anilino]butanoic acid
SMILES OC(=O)CCC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2nccs2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H13N3O5S2/c17-11(5-6-12(18)19)15-9-1-3-10(4-2-9)23(20,21)16-13-14-7-8-22-13/h1-4,7-8H,5-6H2,(H,14,16)(H,15,17)(H,18,19)
Formula Weight 355.0297
Chemical Name or Material Succinylsulfathiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.