missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan Black B, Pure, C.I.26150, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Melting Point | 120°C |
| CAS | 4197-25-5 |
| Molecular Formula | C29H24N6 |
| MDL Number | 6919 |
| InChI Key | YCUVUDODLRLVIC-UHFFFAOYSA-N |
| SMILES | CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C |
| IUPAC Name | (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene |
| Molecular Weight (g/mol) | 456.553 |
| PubChem CID | 61336 |
| Formula Weight | 456.54g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction