missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan Orange G (~85%), TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | Sudan Orange G |
| CAS | 2051-85-6 |
| Molecular Formula | C12 H10 N2 O2 |
| Quantity | 2.5 g |
| Synonym | Sudan Orange G, 2,4-Dihydroxyazobenzene, 1,3-Benzenediol','4-(2-phenyldiazenyl)- (ACI), 1,3-Benzenediol','4-(phenylazo)- (9CI), 4-(2-Phenyldiazenyl)-1,3-benzenediol (ACI), C.I. Solvent Orange 1 (7CI','8CI), Sudan G (6CI), 1504 Yellow, 2,4-Dibenzeneazoresorcinol, 2,4-Dihydroxyazobenzene, 4-(Phenylazo)resorcinol, Benzeneazoresorcinol, C.I. 11920, C.I. Food Orange 3, Ceres Orange, Ceres Orange G, Ceres orange GN, Cerisol Yellow GR, Fast Oil Orange T, Fast Oil Yellow 2G, Fast Oil Yellow G, Fat Orange A, Fat Orange G, Fat Orange GS, Fat Orange RG, Fat Victoria Yellow D, Grasol Yellow RSF, Hexacol Oil Yellow GG, Lacquer Orange V 3G, NSC 7949, Oil Orange 4G, Oil Orange G, Oil Orange MO, Oil Orange MON, Oil Orange MON Extra, Oil Yellow G Extra, Oil Yellow GG, Oil-sol. Yellow Zh, Orange Au Gras W 2201, Organol Orange 2J, Plastoresin Orange F 3A, Resinol Orange G, Solvent Orange 1, Sudan Orange G, Sudan Yellow AR, Tertrogras Orange SG, Yellow M Soluble in Grease |
| InChI Formula | InChI=1S/C12H10N2O2/c15-10-6-7-11(12(16)8-10)14-13-9-4-2-1-3-5-9/h1-8,15-16H |
| SMILES | Oc1ccc(N=Nc2ccccc2)c(O)c1 |
| IUPAC Name | 4-[(E)-phenyldiazenyl]benzene-1,3-diol |
| Molecular Weight (g/mol) | 214.22 |
| Formula Weight | 214.0742 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction