missing translation for 'onlineSavingsMsg'
Learn More

Sudan Orange G, TRC

Product Code. 30426508
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30426508

Brand: Toronto Research Chemicals TRCS688925100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sudan Orange G
CAS 2051-85-6
Molecular Formula C12 H10 N2 O2
Quantity 100 mg
Synonym Sudan Orange G, 2,4-Dihydroxyazobenzene, 1,3-Benzenediol','4-(2-phenyldiazenyl)- (ACI), 1,3-Benzenediol','4-(phenylazo)- (9CI), 4-(2-Phenyldiazenyl)-1,3-benzenediol (ACI), C.I. Solvent Orange 1 (7CI','8CI), Sudan G (6CI), 1504 Yellow, 2,4-Dibenzeneazoresorcinol, 2,4-Dihydroxyazobenzene, 4-(Phenylazo)resorcinol, Benzeneazoresorcinol, C.I. 11920, C.I. Food Orange 3, Ceres Orange, Ceres Orange G, Ceres orange GN, Cerisol Yellow GR, Fast Oil Orange T, Fast Oil Yellow 2G, Fast Oil Yellow G, Fat Orange A, Fat Orange G, Fat Orange GS, Fat Orange RG, Fat Victoria Yellow D, Grasol Yellow RSF, Hexacol Oil Yellow GG, Lacquer Orange V 3G, NSC 7949, Oil Orange 4G, Oil Orange G, Oil Orange MO, Oil Orange MON, Oil Orange MON Extra, Oil Yellow G Extra, Oil Yellow GG, Oil-sol. Yellow Zh, Orange Au Gras W 2201, Organol Orange 2J, Plastoresin Orange F 3A, Resinol Orange G, Solvent Orange 1, Sudan Orange G, Sudan Yellow AR, Tertrogras Orange SG, Yellow M Soluble in Grease
InChI Formula InChI=1S/C12H10N2O2/c15-10-6-7-11(12(16)8-10)14-13-9-4-2-1-3-5-9/h1-8,15-16H
SMILES Oc1ccc(N=Nc2ccccc2)c(O)c1
IUPAC Name 4-[(E)-phenyldiazenyl]benzene-1,3-diol
Molecular Weight (g/mol) 214.22
Formula Weight 214.0742
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.