missing translation for 'onlineSavingsMsg'
Learn More

Sudan Orange G, TRC

Product Code. 30410322
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30410322

Brand: Toronto Research Chemicals TRCS6889251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sudan Orange G
CAS 2051-85-6
Molecular Formula C12 H10 N2 O2
Quantity 1 g
Synonym Sudan Orange G, 2,4-Dihydroxyazobenzene, 1,3-Benzenediol','4-(2-phenyldiazenyl)- (ACI), 1,3-Benzenediol','4-(phenylazo)- (9CI), 4-(2-Phenyldiazenyl)-1,3-benzenediol (ACI), C.I. Solvent Orange 1 (7CI','8CI), Sudan G (6CI), 1504 Yellow, 2,4-Dibenzeneazoresorcinol, 2,4-Dihydroxyazobenzene, 4-(Phenylazo)resorcinol, Benzeneazoresorcinol, C.I. 11920, C.I. Food Orange 3, Ceres Orange, Ceres Orange G, Ceres orange GN, Cerisol Yellow GR, Fast Oil Orange T, Fast Oil Yellow 2G, Fast Oil Yellow G, Fat Orange A, Fat Orange G, Fat Orange GS, Fat Orange RG, Fat Victoria Yellow D, Grasol Yellow RSF, Hexacol Oil Yellow GG, Lacquer Orange V 3G, NSC 7949, Oil Orange 4G, Oil Orange G, Oil Orange MO, Oil Orange MON, Oil Orange MON Extra, Oil Yellow G Extra, Oil Yellow GG, Oil-sol. Yellow Zh, Orange Au Gras W 2201, Organol Orange 2J, Plastoresin Orange F 3A, Resinol Orange G, Solvent Orange 1, Sudan Orange G, Sudan Yellow AR, Tertrogras Orange SG, Yellow M Soluble in Grease
InChI Formula InChI=1S/C12H10N2O2/c15-10-6-7-11(12(16)8-10)14-13-9-4-2-1-3-5-9/h1-8,15-16H
SMILES Oc1ccc(N=Nc2ccccc2)c(O)c1
IUPAC Name 4-[(E)-phenyldiazenyl]benzene-1,3-diol
Molecular Weight (g/mol) 214.22
Formula Weight 214.0742
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado