missing translation for 'onlineSavingsMsg'
Learn More

Sulfabenzamide, 98.5%, Thermo Scientific™

Product Code. 10665462 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665462 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10665462 Supplier Thermo Scientific Acros Supplier No. 235350500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127-71-9
Melting Point 180.0°C to 184.0°C
Color White
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.50%
Packaging Glass bottle
Molecular Formula C13H12N2O3S
MDL Number MFCD00044890
Quantity 50 g
Merck Index 15, 9029
Synonym sulfabenzamide, sulfabenzide, sulfabenzid, sulfabenzoylamide, n-sulfanilylbenzamide, n-sulfamylbenzamide, n-benzoylsulfanilamide, sulfamylbenzamide, sulfabenzamid, sulfabenzamida
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: ca. 16g/100mL 1n naoh or 1n koh,1g/33mL 95% ethanol,1g/9mL acetone,practically insoluble in toluene
InChI Key PBCZLFBEBARBBI-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)NS(=O)(=O)C2=CC=C(C=C2)N
IUPAC Name N-(4-aminophenyl)sulfonylbenzamide
Molecular Weight (g/mol) 276.3
PubChem CID 5319
Formula Weight 276.3
Percent Purity 98.5%
Physical Form Powder
Chemical Name or Material Sulfabenzamide, 98.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.