Learn More
Sulfadiazine-d4, TRC
Product Code. 30409830
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409830 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30409830 Supplier Toronto Research Chemicals Supplier No. TRCS6990511MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfadiazine D4
CAS 1020719-78-1
Purity Grade Notes HPLC
Molecular Formula C10 D4 H6 N4 O2 S
Quantity 1 mg
Synonym 4-Amino-N-2-pyrimidinylbenzene-2,3,5,6-d4-sulfonamide, Benzene-2,3,5,6-d4-sulfonamide','4-amino-N-2-pyrimidinyl-, Sulfadiazine D4
InChI Formula InChI=1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2ncccn2
IUPAC Name 4-amino-2,3,5,6-tetradeuterio-N-pyrimidin-2-ylbenzenesulfonamide
Molecular Weight (g/mol) 254.302
Formula Weight 254.078
Percent Purity >95