Learn More
Sulfadoxine, LoGiCal
Product Code. 30463919
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463919 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30463919 Supplier LoGiCal Supplier No. LGCFOR0533.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sulfadoxine
CAS 2447-57-6
Analyte or Component Names Sulfadoxine
Shipping Condition Room Temperature
Molecular Formula C12 H14 N4 O4 S
Certifications/Compliance ISO 9001
Synonym Sulfadoxine, 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(5,6-dimethoxy-4-pyrimidinyl)-, 4,5-Dimethoxy-6-sulfanilamidopyrimidine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzensulfonamide, 4-Sulfanilamido-5,6-dimethoxypyrimidine, Fanasil, Fanasulf, N1-5,6-Dimethoxy-4-pyrimidinylsulfanilamide, Orthosulfin, Ro 4-4393, Sanasil, Sulfadoxin, Sulformethoxine, Sulformetoxin, Sulformetoxine, Sulforthodimethoxine, Sulforthomidine, WR 4073
InChI Formula InChI=1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)
Solution Type Neat
SMILES COc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC
IUPAC Name 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 310.33
Quantity 10 mg
Formula Weight 310.0736 g/mol