missing translation for 'onlineSavingsMsg'
Learn More

Sulfameter, TRC

Product Code. 30421270
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421270 100 mg 100mg
30411426 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421270 Supplier Toronto Research Chemicals Supplier No. TRCS699045100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 651-06-9
Molecular Formula C11 H12 N4 O3 S
Molecular Weight (g/mol) 280.3
Synonym Sulfameter
IUPAC Name 4-amino-N-(5-methoxypyrimidin-2-yl)benzenesulfonamide
SMILES COc1cnc(NS(=O)(=O)c2ccc(N)cc2)nc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H12N4O3S/c1-18-9-6-13-11(14-7-9)15-19(16,17)10-4-2-8(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)
Formula Weight 280.063
Chemical Name or Material Sulfameter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.