missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxazole, 98%

Product Code. 15487969
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15487969 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15487969 Supplier Thermo Scientific Alfa Aesar Supplier No. J66565.22
  • ONLINE EXCLUSIVE PRICE
    204.00€
    183.65€ / 100g
    Save 20.35€
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sulfamethoxazole, CAS # 723-46-6, is a sulfonamide antibiotic that inhibits folic acid synthesis in bacteria and prevents cell division and growth. | CAS: 723-46-6 | C10H11N3O3S | 253.276 g/mol

A broad spectrum bacteriostatic sulfonamide antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sulfamethoxazole is an antibiotic composed of an isoxazole with a 4-aminobenzenesulfonamido and methyl-group substitution at positions 5 and 3, respectively
  • Sulfamethoxazole carries out most of its action through the inhibition of dihydropteroate synthase, the enzyme responsible for the synthesis of the precursor of folic acid; inhibition of folic acid synthesis prevents DNA replication and cell division in susceptible bacteria

Application

  • Sulfamethoxazole can elicit an allergic response and may be useful for immune cell studies in laboratory settings
  • Sulfamethoxazole is suitable for laboratory research on infection, environmental pollution, and the physiology of susceptible bacteria
TRUSTED_SUSTAINABILITY

Specifications

CAS 723-46-6
Melting Point 168°C to 172°C
Boiling Point 482°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Molecular Formula C10H11N3O3S
MDL Number MFCD00010546
Quantity 100 g
Beilstein 6732984
Sensitivity Light sensitive
Synonym sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
Solubility Information Soluble in ethanol or acetone. Very slightly soluble in water
InChI Key JLKIGFTWXXRPMT-UHFFFAOYSA-N
SMILES CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N
IUPAC Name 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
Molecular Weight (g/mol) 253.276
PubChem CID 5329
ChEBI CHEBI:9332
Formula Weight 253.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulfamethoxazole
Show More Show Less
compliance-icons
Product Identifier
  • Sulphamethoxazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sulphamethoxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.