missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxazole, TRC

Product Code. 17974865
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17974865 50 mg 50mg
17901315 1 g 1g
17953025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974865 Supplier Toronto Research Chemicals Supplier No. TRCS69908650MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723-46-6
Molecular Formula C10 H11 N3 O3 S
Molecular Weight (g/mol) 253.28
Synonym Sulfamethoxazole, 4-Amino-N-(5-methylisoxazol-3-yl)benzenesulfonamide
IUPAC Name 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
SMILES Cc1onc(NS(=O)(=O)c2ccc(N)cc2)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H11N3O3S/c1-7-6-10(12-16-7)13-17(14,15)9-4-2-8(11)3-5-9/h2-6H,11H2,1H3,(H,12,13)
Formula Weight 253.0521
Percent Purity >95%
Chemical Name or Material Sulfamethoxazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.